missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Cr/Mo, Alloy C-22, N06022, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Cr/Mo, Alloy C-22, N06022, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Oxygen/Carbon/Titanium/Magnesium/Phosphorus/Vanadium/Nitrogen/Niobium/Copper/Manganese/Aluminium/Cobalt/Tungsten/Iron/Molybdenum/Chromium/Nickel/Boron/Tin/Zirconium/Tantalum |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.0005 %/Carbon 0.0021 %/Titanium 0.005 %/Magnesium 0.007 %/Phosphorus 0.008 %/Vanadium 0.012 %/Nitrogen 0.019 %/Niobium 0.033 %/Copper 0.05 %/Manganese 0.28 %/Aluminium 0.29 %/Cobalt 1.22 %/Tungsten 2.81 %/Iron 3.66 %/Molybdenum 13.1 %/Chromium 21.5 %/Nickel 56.8 %/Boron (0.001) %/Tin (0.001) %/Zirconium (0.002) %/Tantalum (0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction