missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGB 2904, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
NGB 2904 is a potent and selective dopamine D3 receptor antagonist (Ki values are 1.4, 217, 223, 642, > 5000, > 10000 and > 10000 nM for D3, D2, 5-HT2, α1, D4, D1 and D5 receptors respectively). Potently antagonizes quinpirole -stimulated mitogenesis (IC50 = 6.8 nM). Attenuates cocaine's rewarding effects and inhibits relapse to drug-seeking behavior.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 530.92 |
Percent Purity | >98% |
Chemical Name or Material | NGB 2904 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'