Learn More
LNI Swissgas NG CASTORE 3G 45
Product Code. 17812169 Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17812169
Brand: LNI Swissgas 6920.51.045
Regulatory: This product is under eco-contribution tax.
Nitrogen generator with 2 air outlet (no dryer).
  • Needs source of dry and oil free air
  • For Sciex instruments
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator