missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFYC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565917
This item is not returnable.
View return policy
Description
Immunogen sequence: EPVQYYFTLAQ QPTAVQVQGQ QQGQQTTSST TTIQPGQIII AQPQQGQTTP VTMQVGEGQQ VQIVQAQPQG QAQQAQSGTG QTMQVMQQII TNTGEIQQIP VQLNAGQLQY IRLAQPVSGT QVVQGQIQTL ATNAQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.
The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein histone-fold "e;handshake"e; motif in a manner analogous to the histone proteins, H2B and H2A, respectively.Specifications
NFYC | |
Polyclonal | |
Unconjugated | |
NFYC | |
CAAT box DNA-binding protein subunit C; CAAT-box DNA-binding protein subunit C; CBFC; CBF-C; CCAAT binding factor of CBF-C/NFY-C; CCAAT binding factor subunit C; CCAAT transcription binding factor subunit gamma; CCAAT-binding transcription factor subunit C; DKFZp667G242; fb98f01; FLJ45775; H1TF2A; HAP5; histone H1 transcription factor large subunit 2A; HSM; NFYC; NF-YC; Nuclear transcription factor Y subunit C; nuclear transcription factor Y subunit gamma; nuclear transcription factor Y, gamma; nuclear transcription factor-Y gamma; transactivator HSM-1; transactivator HSM-1/2; transcription factor NF-Y, C subunit; wu:fb98f01; zgc:55514 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13952 | |
NFYC | |
Recombinant Human NFYC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction