Learn More
NFkB p105/p50, p Ser337 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-82074-0.025ML
 View more versions of this product
Product Code. 18281903
  • 258.00€ / 25µL
Description
NFkB p105/p50 Polyclonal specifically detects NFkB p105/p50 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFkB p105/p50 (p Ser337)
Polyclonal
Unconjugated
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol with 0.02% Sodium Azide
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
Rabbit
Affinity Purified
RUO
Primary
NF-kB p105/p50 (specific to Phospho-Ser337) detects endogenous levels of NF-kB p105/p50 only when phosphorylated at serine 337.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P19838
NFKB1
The antiserum was produced against synthesized phosphopeptide derived from human NF-kB p105/p50 around the phosphorylation site of serine 337 (R-K-SP-D-L).
0.025 mL
Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
4790
Human, Rat, Mouse