Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Tubes
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette.
168.00€ - 660.00€
Specifications
For Use With (Equipment) | Neon™ NxT Electroporation System |
---|---|
For Use With (Application) | Electroporation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18065321
|
Invitrogen™
NT96 |
96 tubes |
660.00€
Pack of 96 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30245030
|
Invitrogen™
NT24 |
24 Tubes |
168.00€
Pack of 24 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight 1-channel tubes, and each Neon NxT Electroporation System Kit with 1-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 1-channel tubes. This pack of additional tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup or for reactions with the 1-channel pipette when predominately using the 8-channel pipette and utilizing kits that contain the 8-channel tubes.
Specifications
Neon™ NxT Electroporation System | |
Electroporation |
For Research Use Only. Not for use in diagnostic procedures.