Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Pipette Station
The Neon NxT Electroporation System Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1).
Brand: Invitrogen™ NEON1PS
Description
The Neon NxT Electroporation System 1-Channel Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1S). Each Neon NxT Electroporation System (Cat. Nos. NEON1S, NEON1SK, NEON18S and NEON18SK) comes with a 1-channel pipette station. This 1-channel pipette station is offered separately for those needing an additional pipette station or replacement for their existing pipette station.
Specifications
Neon™ NxT 10-μL and 100-μL tips | |
Transfection | |
1 station |
Electroporation Pipette Station | |
Neon™ NxT Electroporation System |
For Research Use Only. Not for use in diagnostic procedures.