Learn More
Thermo Scientific™ Neisseria Selective Agar
Isolate and cultivate pathogenic Neisseria species with a special formulation, including a peptone mixture of meat and plant enzymatic digests, starch and phosphate buffers.
Brand:  Thermo Scientific™ PO5004A
Product Code. 13225349
  • 14.65€ / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Isolate and cultivate pathogenic Neisseria species with Thermo Scientific™ Neisseria Selective Agar, a medium specially formulated with a peptone mixture that includes meat and plant enzymatic digests and the presence of starch to ensure that toxic metabolites produced by Neisseria are absorbed. Phosphate buffers are also included to prevent changes in pH due to amine production that would affect the survival of the organism.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tested in accordance with CEN ISO/TS 11133-2. Formulation of this medium conforms to ISO 4833.
6°C to 12°C
90mm Monoplate
Agar
Flow Wrapped - Single Layer
Signal brown, opaque
Neisseria Selective Agar
Germany
17g ± 5%
10/Pk.