Learn More
BTX™ Needles Array Electrode Handles for BTX™ AgilePulse™ in Vivo
Needle Array Handle for miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ in vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
2140.00€
Specifications
For Use With (Application) | Gene and Vaccine Delivery |
---|---|
Material | Plastic |
Product Line | BTX Electroporation |
Product Type | Needle Array Handle |
Includes | Handle Only |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15417370
|
BTX™
|
Generators with SN AP-4068-ID/AP-5030-IM and Before |
2140.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
Gene and Vaccine Delivery | |
BTX Electroporation | |
Handle Only | |
Non-sterile | |
Square Wave |
Plastic | |
Needle Array Handle | |
1 Ea. | |
Electrode Connector Cable |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.