missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nebulin Antibody (NB2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-11099
This item is not returnable.
View return policy
Description
Nebulin Monoclonal specifically detects Nebulin in Human, Mouse, Rat, Porcine, Alligator, Amphibian, Avian, Bovine, Chicken, Fish, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Nebulin | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 15mM Sodium Azide | |
| NEB | |
| Full length native protein (purified) (Chicken). | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 4703 | |
| Human, Mouse, Rat, Pig, Alligator, Amphibian, Avian, Bovine, Chicken, Fish, Rabbit | |
| Ascites |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| NB2 | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:400 | |
| DKFZp686C1456, FLJ11505, FLJ36536, FLJ39568, FLJ39584, NEB177D, nebulin, NEM2 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Localizes nebulin in skeletal muscle of a wide variety of species from cold-blooded vertebrates to man. Does not crossreact with smooth or cardiac muscle, non-muscle tissue and cultured cells. Localizes nebulin within the I band of skeletal muscle sections by indirect immunofluorescence. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction