Learn More
Abeba™ Nature 4080 Shoes
Leather covered cork footbed is ESD dissipative.
84.65€
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC |
---|---|
Color | White |
Footbed Material | Cork |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15621428
|
Abeba™
4080/34 |
34 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15631428
|
Abeba™
4080/35 |
35 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15641428
|
Abeba™
4080/36 |
36 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15651428
|
Abeba™
4080/37 |
37 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15661428
|
Abeba™
4080/38 |
38 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15671428
|
Abeba™
4080/39 |
39 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15681428
|
Abeba™
4080/40 |
40 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15691428
|
Abeba™
4080/41 |
41 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15601438
|
Abeba™
4080/42 |
42 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15611438
|
Abeba™
4080/43 |
43 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15621438
|
Abeba™
4080/44 |
44 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15631438
|
Abeba™
4080/45 |
45 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15641438
|
Abeba™
4080/46 |
46 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15651438
|
Abeba™
4080/47 |
47 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15661438
|
Abeba™
4080/48 |
48 |
84.65€
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- 2 adjustable instep straps
- velcro fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
Slip-resistant | |
4080 |
White | |
Leather | |
Rubber |