Learn More
Piercan™ Natural Rubber Latex Chemical Resistant Gloves
Product Code. 11799528
Click to view available options
Size:
8
Specifications
Certifications/Compliance ISO 9001 and ISO 14001
Coating Material Natural Rubber Latex
Hand Ambidextrous
Latex Free Yes
Material Rubber Latex
Product Type Gloves
Size 8
Thickness (Metric) 0.4 mm
Quantity 1 Pair
Resistant To Alkalis, Aliphatic solvents and Ketones
Length (Metric) 800 mm