Learn More
Fisherbrand™ Natural Polypropylene Centrifuge Tube
Attach the Fisherbrand™ Natural Polypropylene Centrifuge Tube to a laboratory centrifuge for the simple and safe processing of samples.
Brand:  Fisherbrand™ 471A6
Product Code. 11951869
  • 345.56€ / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • 50mL capacity
  • 3000xg Max force and Max RCF capacities
  • Sterile
Specifications
Specifications
Natural
Polypropylene
500
Conical
Centrifuge Tube
50 mL
Centrifugation
Sterile
Conical
3000 x g