Learn More
Thermo Scientific™ National Snap-It™ Closures for 12 x 32 crimp/snap vials
Product Code. 12493768
Click to view available options
Color:
Clear
Green
Yellow
Septum:
Integral Polyethylene
Integral Star Slit
PTFE/Silicone/PTFE
Viton™ Elastomer
Unit Size:
Pack of 100
Pack of 1000
Product Code. 12493768
Brand: Thermo Scientific™ C401157
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
TRUSTED_SUSTAINABILITY
Specifications
Description C4011-57
Color Clear
Pre-slit Yes
Volume (Metric) 2 mL
Septum Integral Star Slit
Product Line National Snap-It
Closure Size 11 mm