Learn More
Thermo Scientific™ National Crimp-Top Autosampler Vials
Product Code. 10702357 Shop All Thermo Scientific Products
Click to view available options
Description:
2.5mL; 11mm; Clear
8mm x 40mm crimp top vials, clear glass flat bottom, 1.2mL
Volume (Metric):
1.2 mL
2.5 mL
Unit Size:
Pack of 100
Pack of 125
Product Code. 10702357
Brand: Thermo Scientific™ 60180503
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Crimp-Top Autosampler Vials are offered as clear or amber, in a range of sizes.
Thermo Scientific™ National Crimp-Top Autosampler Vials are offered as clear or amber, in a range of sizes.
Ordering Information:
Order caps separately.
TRUSTED_SUSTAINABILITY
Specifications
Description 2.5mL; 11mm; Clear
Volume (Metric) 2.5 mL
Volume (Metric) Working 2.5 mL
Closure Size 11 mm