Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Product Code. 11985673
Click to view available options
Color:
Amber
Clear
Closure Type:
Phenolic
Polypropylene
Polypropylene, Flanged
Septum:
PTFE
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Packaging Type:
Polybag
Storage Tray
Unit Size:
Pack of 100
Product Code. 11985673
Brand: Thermo Scientific™ C4013195
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color (Seal) Red/White
Description Clear Glass, flanged; PP Black Cap; Red PTFE/White Silicone Preslit septum; in storage tray
Color Clear
Material Glass, PTFE, Silicone
Pre-slit Yes
Volume (Metric) 2 mL
Septum PTFE/Silicone, Pre-slit
Packaging Type Storage Tray
Closure Material Polypropylene
Closure Type Polypropylene, Flanged
Closure Size 8-425