missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-thiocarboxamide, 97%, Thermo Scientific™

Product Code. 15468777
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20300-10-1
Molecular Formula C11H9NS
Molecular Weight (g/mol) 187.26
MDL Number MFCD08164114
InChI Key DRCKUACWKCMOCB-UHFFFAOYSA-N
Synonym 1-naphthalenecarbothioamide, naphthalene-1-thiocarboxamide
PubChem CID 3981658
IUPAC Name naphthalene-1-carbothioamide
SMILES C1=CC=C2C(=C1)C=CC=C2C(=S)N

Specifications

Quantity 1 g
Formula Weight 187.26
Percent Purity 97%
Chemical Name or Material Naphthalene-1-thiocarboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'