missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nanaocolor™ Ortho- and Total Phosphate 15 Tube Tests

Guarantees maximum safety for the user because measurement is performed directly in the test tube. Macherey Nagel™ Nanocolor™ Ortho- and Total Phosphate 15 Tube Tests automatically perform the measurement after the cuvette is inserted into the cuvette slot.

Brand:  Macherey-Nagel™ 985080

Additional Details : Weight : 1.00000kg


Product Code. 10774673

  • 96.75€ / 1 test
In Stock
to see stock.
Add to basket

Description

Description

  • Ammonium molybdate reacts with orthophosphate ions to form phosphomolybdic acid
  • Sea water suitability
  • For the determination of total phosphate, an acidic oxidation at 100-120°C must precede to detect poly- and organic phosphates
  • Photometric determination of phosphate
  • Reaction basis based on international approved standard methods like DIN-, EN-, ISO-, EPA- and APHA
  • Tests contain accurate predosed reagents that are directly ready to use
  • Every test box is equipped with a color-coded label and can be identified clearly
Specifications

Specifications

20 Determinations
0.30 - 15.00mg/L P (PO4-P)
1.0 - 45.0mg/L PO43-
tube test 0-80
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • NANOCOLOR ortho and total Phosphate 15

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : sodium peroxodisulfate, sulfuric acid

SDS
Documents

Documents

Certificates
Promotions

Promotions