missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Fmoc-L-lysine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.
Solubility
Soluble in dimethyl formamide (0.3g in 2ml).
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.
Solubility
Soluble in dimethyl formamide (0.3g in 2ml).
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Beilstein | 8663370 |
Solubility Information | Soluble in dimethyl formamide (0.3g in 2ml). |
Formula Weight | 404.89 |
Percent Purity | 98% |
Chemical Name or Material | Nalpha-Fmoc-L-lysine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'