Learn More
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
GREENER_CHOICE
Product Code. p-4525018
Click to view available options
Diameter (Metric)Top:
35 mm
54 mm
63 mm
76 mm
98 mm
Capacity (Metric):
10 mL
41 mL
64 mL
114 mL
254 mL
Unit Size:
Pack of 12
Pack of 4
Pack of 6
Product Code. 10010741
Brand: Thermo Scientific™ 42500035
Only null left
Add to basket
Add to basket
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 35 mm
Capacity (Metric) 10 mL
Diameter (Metric) Stem 6 mm
Material Polypropylene
Autoclavable Autoclavable
Description Funnel Analytical PP 35mm
Length (Metric) Stem 50 mm
Quantity 12/Pk.; 36/Cs.