Learn More
NaDC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26184-1-AP-20UL
 View more versions of this product
Product Code. 16864705
  • 145.00€ / 20µL
Description
Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
Specifications
Specifications
NaDC3
Polyclonal
Unconjugated
SLC13A3
hNaDC3, NaDC 3, NADC3, SDCT2, SLC13A3
Rabbit
Antigen Affinity Chromatography
RUO
114644, 64849
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin)
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WWT9, Q91Y63
SLC13A3
NaDC3 Fusion Protein Ag24182
20 μL
Primary
Mouse, Human