missing translation for 'onlineSavingsMsg'
Learn More

N1-(3-acetyl-4-hydroxyphenyl)acetamide, Thermo Scientific™

Product Code. p-4391143
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10096412

Brand: Thermo Scientific Maybridge BTB10078DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7298-67-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key DIQSYMRVTOVKQT-UHFFFAOYSA-N
Synonym n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
PubChem CID 81720
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=CC(=C1)NC(=O)C)O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material N1-(3-Acetyl-4-hydroxyphenyl)acetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.