missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylglycine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 15421389
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-23-7
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD03788441
InChI Key QORBVLNEGUIPND-UHFFFAOYSA-N
Synonym 2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
PubChem CID 17748195
IUPAC Name hydrogen 2-(tert-butylamino)acetic acid chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)=O

Specifications

Melting Point 223°C to 224°C
Color White
Linear Formula (CH3)3CNHCH2COOH·HCl
Beilstein 6296616
Formula Weight 167.63
Percent Purity 97%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-tert-Butylglycine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'