missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diaminoguanidine monohydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10521801
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36062-19-8
Molecular Formula CH7N5·HCl
Molecular Weight (g/mol) 125.56
MDL Number MFCD00012948
InChI Key HAZRIBSLCUYMQP-UHFFFAOYSA-N
Synonym 1,3-diaminoguanidine hydrochloride, unii-014y506oeg, 1,3-diaminoguanidine monohydrochloride, carbonimidic dihydrazide, hydrochloride, n,n'-diaminoguanidine monohydrochloride, n,n'-diaminoguanidine, carbonimidic dihydrazide, hydrochloride 1:?, 1,2-diaminoguanidine hydrochloride, n,n'-diaminoguandine, hydrochloride, pubchem20888
PubChem CID 9566041
IUPAC Name 1,2-diaminoguanidine;hydrochloride
SMILES C(=NN)(N)NN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 178°C to 182°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NNHC(=NH)NHNH2·HCl
Quantity 25 g
Beilstein 03, 122
Formula Weight 125.56
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material N, N'-Diaminoguanidine monohydrochloride, 98%
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.