missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-trimethylsilylacetamide, 98%

Product Code. 11366786
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11366786

Brand: Thermo Scientific Alfa Aesar L04101.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7449-74-3 | C6H15NOSi | 145.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7449-74-3
Molecular Formula C6H15NOSi
Molecular Weight (g/mol) 145.277
MDL Number MFCD00040758
InChI Key QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonym n-methyl-n-trimethylsilyl acetamide, acetamide, n-methyl-n-trimethylsilyl, n-trimethylsilyl-n-methylacetamide, n-methyl-n-tms-acetamide, acmc-1bei6, qhuobldkfgcvcg-uhfffaoysa, n-methylacetamide, tms derivative, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
PubChem CID 81953
IUPAC Name N-methyl-N-trimethylsilylacetamide
SMILES CC(=O)N(C)[Si](C)(C)C

Specifications

Boiling Point 48°C to 49°C (11mmHg)
Quantity 2 g
UN Number UN1993
Beilstein 506975
Flash Point 48°C (118°F)
Formula Weight 145.28
Percent Purity 95%
Refractive Index 1.439
Sensitivity Moisture sensitive
Density 0.906
Chemical Name or Material N-Methyl-N-trimethylsilylacetamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.