missing translation for 'onlineSavingsMsg'
Learn More

n-Hexylsulfonylacetonitrile, 98%, Thermo Scientific™

Product Code. 11312876
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 203310-42-3
Molecular Formula C8H15NO2S
Molecular Weight (g/mol) 189.27
MDL Number MFCD00041475
InChI Key CGKSNJYMJMDPQT-UHFFFAOYSA-N
Synonym 2-hexylsulfonyl acetonitrile, n-hexylsulfonylacetonitrile, n-hexanesulphonylacetonitrile, hexane-1-sulfonylacetonitrile, 2-hexane-1-sulfonyl acetonitrile
PubChem CID 2817563
IUPAC Name 2-hexylsulfonylacetonitrile
SMILES CCCCCCS(=O)(=O)CC#N

Spezifikation

Melting Point 41°C to 43°C
Quantity 1 g
Formula Weight 189.28
Percent Purity 98%
Chemical Name or Material n-Hexylsulfonylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'