missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, Riedel-de Haën™

Product Code. 15975470
Click to view available options
Quantity:
7 L
Packaging:
Returnable Stainless Steel Can
Grade:
GC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

For pesticide residue analysis, for analysis of dioxins, furans and PCB

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -95°C
CAS 110-54-3
Color Colorless
Boiling Point 66°C to 69°C (1.013 hPa)
Quantity 7 L
Packaging Returnable Stainless Steel Can
Molecular Formula C6H14
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES CCCCCC
IUPAC Name hexane
Molecular Weight (g/mol) 86.18
Grade GC
Chemical Name or Material n-Hexane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'