Learn More
N-Fmoc-L-glutamic acid, 95%, Thermo Scientific Chemicals
39.70€ - 520.00€
Chemical Identifiers
CAS | 121343-82-6 |
---|---|
Molecular Formula | C20H19NO6 |
Molecular Weight (g/mol) | 369.373 |
MDL Number | MFCD00237657 |
InChI Key | QEPWHIXHJNNGLU-KRWDZBQOSA-N |
Synonym | fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid |
PubChem CID | 7019018 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489378
|
Thermo Scientific Alfa Aesar
H62936.06 |
5 g |
39.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15499378
|
Thermo Scientific Alfa Aesar
H62936.14 |
25 g |
146.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409388
|
Thermo Scientific Alfa Aesar
H62936.22 |
100 g |
520.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySlightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
121343-82-6 | |
369.373 | |
QEPWHIXHJNNGLU-KRWDZBQOSA-N | |
7019018 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
C20H19NO6 | |
MFCD00237657 | |
fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
Specifications
121343-82-6 | |
MFCD00237657 | |
fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid | |
QEPWHIXHJNNGLU-KRWDZBQOSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid | |
7019018 | |
95% |
C20H19NO6 | |
3633624 | |
Slightly soluble in water. | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O | |
369.373 | |
369.37 | |
N-Fmoc-L-glutamic acid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only