missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-aspartic acid 4-benzyl ester, 97%, Thermo Scientific Chemicals
93.00€ - 331.00€
Chemical Identifiers
CAS | 86060-84-6 |
---|---|
Molecular Formula | C26H23NO6 |
Molecular Weight (g/mol) | 445.47 |
MDL Number | MFCD00065630 |
InChI Key | OQGAELAJEGGNKG-UHFFFAOYNA-N |
Synonym | fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh |
PubChem CID | 13966931 |
SMILES | OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478608
|
Thermo Scientific Alfa Aesar
H62011.06 |
5 g |
93.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488608
|
Thermo Scientific Alfa Aesar
H62011.14 |
25 g |
331.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediates.
Solubility
Soluble in methanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
86060-84-6 | |
445.47 | |
OQGAELAJEGGNKG-UHFFFAOYNA-N | |
13966931 |
C26H23NO6 | |
MFCD00065630 | |
fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh | |
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
86060-84-6 | |
C26H23NO6 | |
4609615 | |
Soluble in methanol. | |
OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
445.47 | |
445.46 | |
N-Fmoc-L-aspartic acid 4-benzyl ester |
120°C to 130°C | |
MFCD00065630 | |
fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh | |
OQGAELAJEGGNKG-UHFFFAOYNA-N | |
4-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid | |
13966931 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only