missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-aspartic acid 4-allyl ester, 98%, Thermo Scientific Chemicals
29.30€ - 393.00€
Chemical Identifiers
CAS | 146982-24-3 |
---|---|
Molecular Formula | C22H21NO6 |
Molecular Weight (g/mol) | 395.411 |
MDL Number | MFCD00190874 |
InChI Key | FBNFRRNBFASDKS-IBGZPJMESA-N |
Synonym | fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc |
PubChem CID | 10715692 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
SMILES | C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15470349
|
Thermo Scientific Alfa Aesar
H63585.03 |
1 g |
29.30€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480349
|
Thermo Scientific Alfa Aesar
H63585.06 |
5 g |
95.50€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15490349
|
Thermo Scientific Alfa Aesar
H63585.14 |
25 g |
393.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
146982-24-3 | |
395.411 | |
FBNFRRNBFASDKS-IBGZPJMESA-N | |
10715692 | |
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C22H21NO6 | |
MFCD00190874 | |
fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid |
Specifications
111°C to 115°C | |
C22H21NO6 | |
6670219 | |
Slightly soluble in water. | |
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
395.411 | |
395.41 | |
Solid |
146982-24-3 | |
MFCD00190874 | |
fmoc-asp oall-oh, fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oallyl-oh, fmoc-asp oall oh, fmoc-asp-allyl ester, ambotzfaa1353, fmoc-asp oal-oh, pubchem12449, n-fmoc-l-aspartic acid 4-allyl ester, fmoc-asp oall-oh hplc | |
FBNFRRNBFASDKS-IBGZPJMESA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid | |
10715692 | |
98% | |
N-Fmoc-L-aspartic acid 4-allyl ester |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only