missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
36.50€ - 126.00€
Chemical Identifiers
CAS | 129460-09-9 |
---|---|
Molecular Formula | C23H25NO6 |
Molecular Weight (g/mol) | 411.45 |
MDL Number | MFCD00065631 |
InChI Key | VZXQYACYLGRQJU-UHFFFAOYNA-N |
Synonym | fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu |
PubChem CID | 7017910 |
SMILES | CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488878
|
Thermo Scientific Alfa Aesar
H62355.03 |
1 g |
36.50€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15498878
|
Thermo Scientific Alfa Aesar
H62355.06 |
5 g |
126.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
129460-09-9 | |
411.45 | |
VZXQYACYLGRQJU-UHFFFAOYNA-N | |
7017910 |
C23H25NO6 | |
MFCD00065631 | |
fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu | |
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
129460-09-9 | |
MFCD00065631 | |
fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu | |
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
411.45 | |
411.45 | |
N-Fmoc-L-aspartic acid 1-tert-butyl ester |
C23H25NO6 | |
3658045 | |
VZXQYACYLGRQJU-UHFFFAOYNA-N | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
7017910 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only