Learn More
N-Fmoc-4-methyl-L-phenylalanine, 95%, Thermo Scientific Chemicals
27.80€ - 392.00€
Chemical Identifiers
CAS | 199006-54-7 |
---|---|
Molecular Formula | C25H23NO4 |
Molecular Weight (g/mol) | 401.462 |
MDL Number | MFCD00270197 |
InChI Key | UXLHLZHGQPDMJQ-QHCPKHFHSA-N |
Synonym | fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl |
PubChem CID | 7129826 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid |
SMILES | CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488597
|
Thermo Scientific Alfa Aesar
H52073.MD |
250 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15468597
|
Thermo Scientific Alfa Aesar
H52073.03 |
1 g |
96.80€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478597
|
Thermo Scientific Alfa Aesar
H52073.06 |
5 g |
392.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.
Solubility
Soluble in Dimethylformamide(DMF).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
199006-54-7 | |
401.462 | |
UXLHLZHGQPDMJQ-QHCPKHFHSA-N | |
7129826 | |
CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C25H23NO4 | |
MFCD00270197 | |
fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid |
Specifications
199006-54-7 | |
MFCD00270197 | |
fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl | |
UXLHLZHGQPDMJQ-QHCPKHFHSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid | |
7129826 | |
95% |
C25H23NO4 | |
8799411 | |
Soluble in Dimethylformamide(DMF). | |
CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
401.462 | |
401.46 | |
N-Fmoc-4-methyl-L-phenylalanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only