missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-pyridyl)-D-alanine, 95%

Product Code. 15478757
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15478757

Brand: Thermo Scientific Alfa Aesar H52193.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 205528-30-9 | C23H20N2O4 | 388.42 g/mol

N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205528-30-9
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.42
MDL Number MFCD00672567
InChI Key SCSSXJVRZMQUKA-UHFFFAOYNA-N
Synonym fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid
PubChem CID 978323
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
SMILES OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-pyridyl)-D-alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.