missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Carbobenzyloxy-L-alanine, 98%
26.70€ - 212.00€
Chemical Identifiers
CAS | 1142-20-7 |
---|---|
Molecular Formula | C11H13NO4 |
Molecular Weight (g/mol) | 223.23 |
MDL Number | MFCD00002640 |
InChI Key | TYRGLVWXHJRKMT-QMMMGPOBSA-N |
Synonym | n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine |
PubChem CID | 736104 |
IUPAC Name | (2S)-2-(phenylmethoxycarbonylamino)propanoic acid |
SMILES | C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10543341
|
Thermo Scientific Chemicals
169000050 |
5 g | Glass bottle |
26.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10032230
|
Thermo Scientific Chemicals
169000250 |
25 g | Glass bottle |
76.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299443
|
Thermo Scientific Chemicals
169001000 |
100 g | Glass bottle |
212.00€
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1142-20-7 | |
223.23 | |
TYRGLVWXHJRKMT-QMMMGPOBSA-N | |
736104 | |
C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
C11H13NO4 | |
MFCD00002640 | |
n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine | |
(2S)-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
1142-20-7 | |
100.0 | |
White | |
98% | |
CH3CH(NHCO2CH2C6H5)CO2H | |
-15 | |
Solubility in water: .. Other solubilities: soluble in most common organic solvents | |
C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
223.23 | |
223.23 | |
Powder |
97.5 | |
82°C to 84°C | |
Authentic | |
C11H13NO4 | |
MFCD00002640 | |
n-carbobenzyloxy-l-alanine, z-ala-oh, n-cbz-l-alanine, z-l-alanine, n-benzyloxycarbonyl-l-alanine, cbz-l-alanine, cbz-ala-oh, carbobenzyloxy-l-alanine, benzyloxycarbonyl-l-alanine, carbobenzoxy-l-alanine | |
TYRGLVWXHJRKMT-QMMMGPOBSA-N | |
(2S)-2-(phenylmethoxycarbonylamino)propanoic acid | |
736104 | |
98% | |
N-Carbobenzyloxy-L-alanine |
Safety and Handling
missing translation for 'einecsNumber' : 214-532-8
RUO – Research Use Only