missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-serine, 98% (dry wt.), may cont. up to 10% water, Thermo Scientific Chemicals
22.70€ - 227.00€
Chemical Identifiers
CAS | 3262-72-4 |
---|---|
Molecular Formula | C8H15NO5 |
Molecular Weight (g/mol) | 205.21 |
MDL Number | MFCD00037243 |
InChI Key | FHOAKXBXYSJBGX-MLHKIVSYNA-N |
Synonym | boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser |
PubChem CID | 98766 |
SMILES | CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473327
|
Thermo Scientific Alfa Aesar
A16224.03 |
1 g |
22.70€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483327
|
Thermo Scientific Alfa Aesar
A16224.06 |
5 g |
57.10€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493327
|
Thermo Scientific Alfa Aesar
A16224.14 |
25 g |
227.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3262-72-4 | |
205.21 | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
98766 |
C8H15NO5 | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O |
Specifications
3262-72-4 | |
C8H15NO5 | |
2212252 | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O | |
205.21 | |
205.21 | |
(dry wt.), may cont. up to 10% water |
90°C to 91°C (decomposition) | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
−3.5° (c=2 in Acetic Acid) | |
(2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
98766 | |
98% | |
N-Boc-L-serine |
Safety and Handling
missing translation for 'einecsNumber' : 221-867-3
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only