missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic acid 1,5-dimethyl ester, 97%, Thermo Scientific Chemicals
82.90€ - 341.00€
Chemical Identifiers
CAS | 59279-60-6 |
---|---|
Molecular Formula | C12H21NO6 |
Molecular Weight (g/mol) | 275.301 |
MDL Number | MFCD03094774 |
InChI Key | QNSPKWUAZQIIGZ-QMMMGPOBSA-N |
Synonym | boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester |
PubChem CID | 2734644 |
IUPAC Name | dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate |
SMILES | CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478638
|
Thermo Scientific Alfa Aesar
H62056.06 |
5 g |
82.90€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488638
|
Thermo Scientific Alfa Aesar
H62056.14 |
25 g |
341.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59279-60-6 | |
275.301 | |
QNSPKWUAZQIIGZ-QMMMGPOBSA-N | |
2734644 | |
CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC |
C12H21NO6 | |
MFCD03094774 | |
boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester | |
dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate |
Specifications
59279-60-6 | |
C12H21NO6 | |
boc-glu ome-ome, r-n-boc-glutamic acid-1,5-dimethyl ester, s-dimethyl 2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid dimethyl ester, s-2-tert-butoxycarbonylaminopentanedioic acid dimethyl ester, dimethyl n-tert-butoxycarbonyl-l-glutamate, dimethyl 2s-2-2-methylpropan-2-yl oxycarbonylamino pentanedioate, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, dimethyl ester, 1,5-dimethyl 2s-2-tert-butoxycarbonyl amino pentanedioate, n-boc-l-glutamic acid 1,5-dimethyl ester | |
CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)OC | |
275.301 | |
275.3 | |
Liquid |
Colorless to Yellow | |
MFCD03094774 | |
QNSPKWUAZQIIGZ-QMMMGPOBSA-N | |
dimethyl (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioate | |
2734644 | |
97% | |
N-Boc-L-glutamic acid 1,5-dimethyl ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only