missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-leucine hydrate, 98+%, Thermo Scientific Chemicals
21.20€ - 68.80€
Chemical Identifiers
CAS | 16937-99-8 |
---|---|
Molecular Formula | C11H23NO5 |
Molecular Weight (g/mol) | 249.307 |
MDL Number | MFCD00065583 |
InChI Key | URQQEIOTRWJXBA-DDWIOCJRSA-N |
Synonym | boc-d-leucine monohydrate, r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leu-oh.h2o, n-boc-d-leucine monohydrate, boc-d-leu-oh h2o, d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate, 2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leucine h2o, boc-d-leucine hydrate, n-boc-d-leucinemonohydrate |
PubChem CID | 2724763 |
IUPAC Name | (2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate |
SMILES | CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377236
|
Thermo Scientific Alfa Aesar
L09124.03 |
1 g |
21.20€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387236
|
Thermo Scientific Alfa Aesar
L09124.06 |
5 g |
68.80€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16937-99-8 | |
249.307 | |
URQQEIOTRWJXBA-DDWIOCJRSA-N | |
2724763 | |
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O |
C11H23NO5 | |
MFCD00065583 | |
boc-d-leucine monohydrate, r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leu-oh.h2o, n-boc-d-leucine monohydrate, boc-d-leu-oh h2o, d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate, 2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leucine h2o, boc-d-leucine hydrate, n-boc-d-leucinemonohydrate | |
(2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate |
Specifications
16937-99-8 | |
C11H23NO5 | |
2331060 | |
URQQEIOTRWJXBA-DDWIOCJRSA-N | |
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O | |
249.307 | |
231.29 (Anhydrous) | |
N-Boc-D-leucine hydrate |
80°C to 84°C | |
MFCD00065583 | |
boc-d-leucine monohydrate, r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leu-oh.h2o, n-boc-d-leucine monohydrate, boc-d-leu-oh h2o, d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate, 2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leucine h2o, boc-d-leucine hydrate, n-boc-d-leucinemonohydrate | |
+25° (c=2 in Acetic Acid) | |
(2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate | |
2724763 | |
≥98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only