missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
27.90€ - 85.70€
Chemical Identifiers
CAS | 104719-63-3 |
---|---|
Molecular Formula | C14H25NO6 |
Molecular Weight (g/mol) | 303.355 |
MDL Number | MFCD00076927 |
InChI Key | YGSRAYJBEREVRB-SECBINFHSA-N |
Synonym | boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester |
PubChem CID | 7018748 |
IUPAC Name | (2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
SMILES | CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15459718
|
Thermo Scientific Alfa Aesar
H63172.03 |
1 g |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15469718
|
Thermo Scientific Alfa Aesar
H63172.06 |
5 g |
85.70€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104719-63-3 | |
303.355 | |
YGSRAYJBEREVRB-SECBINFHSA-N | |
7018748 | |
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C |
C14H25NO6 | |
MFCD00076927 | |
boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester | |
(2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
Specifications
104719-63-3 | |
MFCD00076927 | |
boc-d-glu otbu-oh, r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glutamic acid 5-tert-butyl ester, 2r-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu otbu-oh tlc, d-glutamic acid,n-1,1-dimethylethoxy carbonyl-,5-1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C | |
303.355 | |
303.35 | |
Solid |
C14H25NO6 | |
3594191 | |
YGSRAYJBEREVRB-SECBINFHSA-N | |
(2R)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid | |
7018748 | |
98% | |
N-Boc-D-glutamic acid 5-tert-butyl ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only