missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-glutamic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
61.20€ - 804.00€
Chemical Identifiers
CAS | 73872-71-6 |
---|---|
Molecular Formula | C14H25NO6 |
Molecular Weight (g/mol) | 303.36 |
MDL Number | MFCD00076926 |
InChI Key | YMOYURYWGUWMFM-UHFFFAOYNA-N |
Synonym | boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester |
PubChem CID | 7018746 |
SMILES | CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15498848
|
Thermo Scientific Alfa Aesar
H62320.MD |
250 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15478848
|
Thermo Scientific Alfa Aesar
H62320.03 |
1 g |
204.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488848
|
Thermo Scientific Alfa Aesar
H62320.06 |
5 g |
804.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
73872-71-6 | |
303.36 | |
YMOYURYWGUWMFM-UHFFFAOYNA-N | |
7018746 |
C14H25NO6 | |
MFCD00076926 | |
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C |
Specifications
73872-71-6 | |
C14H25NO6 | |
boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester | |
CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C | |
303.36 | |
303.35 | |
Powder |
Brown to White | |
MFCD00076926 | |
YMOYURYWGUWMFM-UHFFFAOYNA-N | |
5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid | |
7018746 | |
95% | |
N-Boc-D-glutamic acid 1-tert-butyl ester |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only