missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-aspartic acid 4-benzyl ester, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 51186-58-4 |
---|---|
Molecular Formula | C16H21NO6 |
Molecular Weight (g/mol) | 323.345 |
MDL Number | MFCD00038255 |
InChI Key | SOHLZANWVLCPHK-GFCCVEGCSA-N |
Synonym | boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester |
PubChem CID | 6994663 |
IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O |
Description
N-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-D-aspartic acid 4-benzyl ester is used as an intermediate in organic synthesis and pharmaceuticals.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
51186-58-4 | |
323.345 | |
SOHLZANWVLCPHK-GFCCVEGCSA-N | |
6994663 | |
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O |
C16H21NO6 | |
MFCD00038255 | |
boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid |
Specifications
51186-58-4 | |
MFCD00038255 | |
boc-d-asp obzl-oh, boc-d-aspartic acid 4-benzyl ester, r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, 2r-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-d-aspartic acid beta-benzyl ester, boc-d-asparticacid4-benzylester, pubchem11179, boc-d-asp obn-oh, boc-d-aspartic acid b-benzyl ester, boc-d-aspartic acid ?-benzyl ester | |
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O | |
323.345 | |
323.34 | |
N-Boc-D-aspartic acid 4-benzyl ester |
C16H21NO6 | |
2305471 | |
SOHLZANWVLCPHK-GFCCVEGCSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid | |
6994663 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only