Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals
52.10€ - 197.00€
Chemical Identifiers
CAS | 3705-42-8 |
---|---|
Molecular Formula | C20H21NO6 |
Molecular Weight (g/mol) | 371.389 |
MDL Number | MFCD00077013 |
InChI Key | VWHKODOUMSMUAF-KRWDZBQOSA-N |
Synonym | z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester |
PubChem CID | 6994031 |
IUPAC Name | (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid |
SMILES | C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2 |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15408066
|
Thermo Scientific Alfa Aesar
H62975.06 |
5 g |
52.10€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418066
|
Thermo Scientific Alfa Aesar
H62975.14 |
25 g |
197.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester used as a potential neuroprotective agent.
Solubility
Soluble in methanol, dimethyl sulfoxide. Slightly soluble in water.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Identificadores de productos químicos
3705-42-8 | |
371.389 | |
VWHKODOUMSMUAF-KRWDZBQOSA-N | |
6994031 | |
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2 |
C20H21NO6 | |
MFCD00077013 | |
z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester | |
(4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid |
Especificaciones
3705-42-8 | |
C20H21NO6 | |
z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester | |
VWHKODOUMSMUAF-KRWDZBQOSA-N | |
(4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid | |
6994031 | |
95% |
98°C to 102°C | |
MFCD00077013 | |
Soluble in methanol,dimethyl sulfoxide. Slightly soluble in water. | |
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2 | |
371.389 | |
371.38 | |
N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester |
Seguridad y manipulación
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only