missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid, 98+%, Thermo Scientific™

Product Code. 11352686
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1152-61-0
Molecular Formula C12H13NO6
Molecular Weight (g/mol) 267.237
MDL Number MFCD00002719
InChI Key XYXYXSKSTZAEJW-VIFPVBQESA-N
Synonym z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
PubChem CID 2723942
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Melting Point 116°C to 121°C
Quantity 5 g
Beilstein 2336722
Optical Rotation +9° (c=2 in Acetic Acid)
Formula Weight 267.24
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'