missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-methoxy-N-phenylbenzamide, 97%, Thermo Scientific™

Product Code. 15425188
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172998-55-9
Molecular Formula C21H19NO2
Molecular Weight (g/mol) 317.388
MDL Number MFCD22683037
InChI Key AAQVKEUFERGCEO-UHFFFAOYSA-N
PubChem CID 84375597
IUPAC Name N-benzyl-2-methoxy-N-phenylbenzamide
SMILES COC1=CC=CC=C1C(=O)N(CC2=CC=CC=C2)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 317.39
Percent Purity 97%
Chemical Name or Material N-Benzyl-2-methoxy-N-phenylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'