missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-alpha-Acetyl-L-glutamine, 99%
Chemical Identifiers
CAS | 2490-97-3 |
---|---|
Molecular Formula | C7H12N2O4 |
Molecular Weight (g/mol) | 188.183 |
MDL Number | MFCD00038159 |
InChI Key | KSMRODHGGIIXDV-YFKPBYRVSA-N |
Synonym | n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine |
PubChem CID | 182230 |
ChEBI | CHEBI:21553 |
IUPAC Name | (2S)-2-acetamido-5-amino-5-oxopentanoic acid |
SMILES | CC(=O)NC(CCC(=O)N)C(=O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2490-97-3 | |
188.183 | |
KSMRODHGGIIXDV-YFKPBYRVSA-N | |
182230 | |
(2S)-2-acetamido-5-amino-5-oxopentanoic acid |
C7H12N2O4 | |
MFCD00038159 | |
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine | |
CHEBI:21553 | |
CC(=O)NC(CCC(=O)N)C(=O)O |
Specifications
∽206°C (decomposition) | |
C7H12N2O4 | |
1727471 | |
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine | |
CC(=O)NC(CCC(=O)N)C(=O)O | |
188.183 | |
CHEBI:21553 | |
99% | |
N-alpha-Acetyl-L-glutamine |
2490-97-3 | |
MFCD00038159 | |
14,25 | |
KSMRODHGGIIXDV-YFKPBYRVSA-N | |
(2S)-2-acetamido-5-amino-5-oxopentanoic acid | |
182230 | |
188.18 | |
−12° (c=3 in Water) |
Safety and Handling
missing translation for 'einecsNumber' : 219-647-7
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only