missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Carboxyphenyl)thiourea, 97%, Thermo Scientific™

Product Code. 11377856
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7366-56-5
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD00041187
InChI Key FOYDMXZTPAEILY-UHFFFAOYSA-N
Synonym 4-thioureidobenzoic acid, 4-carbamothioylamino benzoic acid, 4-2-thioureido benzoic acid, 4-carboxyphenyl thiourea, 1-4-carboxyphenyl-2-thiourea, 1-4-carboxypheny-2-thiourea, 4-thioureido-benzoic acid, n-4-carboxyphenyl thiourea, 4-aminothioxomethyl amino benzoic acid, 4-aminocarbonothioyl amino benzoic acid
PubChem CID 736216
IUPAC Name 4-(carbamothioylamino)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)NC(=S)N

Specifications

Melting Point ∽255°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 2805627
Formula Weight 196.23
Percent Purity 97%
Chemical Name or Material N-(4-Carboxyphenyl)thiourea
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'