missing translation for 'onlineSavingsMsg'
Learn More

N(4)-Acetylcytosine, 98%, Thermo Scientific Chemicals

Product Code. 11365329
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14631-20-0
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00134466
InChI Key IJCKBIINTQEGLY-UHFFFAOYSA-N
PubChem CID 99309
IUPAC Name N-(2-oxo-1H-pyrimidin-6-yl)acetamide
SMILES CC(=O)NC1=CC=NC(=O)N1

Specifications

Melting Point >300°C
Quantity 5 g
Synonym n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material N(4)-Acetylcytosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'