Learn More
Myosin heavy chain 2 Mouse anti-Human, Rat, Rabbit, Clone: TH81 (BGN/04/4481), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65675
This item is not returnable.
View return policy
Description
Myosin heavy chain 2 Monoclonal specifically detects Myosin heavy chain 2 in Human, Rat, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Myosin heavy chain 2 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
| Myosin heavy chain from rabbit muscle | |
| 0.1 mg | |
| Cell Biology, Cellular Markers | |
| 4620 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| ELISA | |
| TH81 (BGN/04/4481) | |
| Unconjugated | |
| PBS (pH 7.2) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |