Learn More
WTW™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meter Set F
Solve multiple measuring tasks at once and save time and costs. WTW7™ MultiLine™ Multi 3630 IDS Multi-Parameter Portable Meters feature three universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
Brand: WTW™ 2FD57F
3977.97 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Three channels, four parameters
- Three measuring channels, freely combinable for the same or different parameters
- For simultaneous multi-parameter measurement without compromise
- Also use the wireless modules together with the new IDS plug head sensors to be independent from cables and measure conveniently at difficult-to-access locations
- Allows the operation with the digital multi-parameter probes MPP 910 IDS and MPP 930 IDS
- Brilliant color graphic display and two USB interfaces
- The batteries can be charged directly in the device
- Archiving user administration can also be used alternatively to mark measuring points
- 10,000 data sets include up to three main parameters, three temperature values as well as the water depth if using the depth probe
Specifications
Yes | |
Sensors: SenTix™ 940, FDO™ 925, TetraCon™ 925 | |
Multi 3630 IDS Set F | |
Multi-parameter portable meter |
3 | |
Manual: 500 records ; Automatic: 10,000 records | |
Wide range power supply, 4 x 1.2 V NiMH battery or via Mini USB-B |