Learn More
Thermo Scientific™ Multidrop Combi nL Reagent Dispensing valve
Optimize dispensing results with this range of accessories.
Brand: Thermo Scientific™ N07494
463.02 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Optimize dispensing results with Thermo Scientific™ Multidrop Combi nL Reagent Dispenser Acccessories.
Specifications
Reagent Dispensing Valve | |
Dispensing valve | |
Each |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92% -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31