missing translation for 'onlineSavingsMsg'
Learn More

Multi-Element Organometallic Oil Standards 23 Element, 100μg/g; SPEX CertiPrep™

Product Code. p-5537516 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 11400359

Brand: SPEX Certiprep S23100Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/g
Al100μg/g
B100μg/g
Ba100μg/g
Ca100μg/g
Cd100μg/g
Cr100μg/g
Cu100μg/g
Fe100μg/g
K100μg/g
Mg100μg/g
Mn100μg/g
Mo100μg/g
Na100μg/g
Ni100μg/g
P100μg/g
Pb100μg/g
Sb100μg/g
Si100μg/g
Sn100μg/g
Ti100μg/g
V100μg/g
Zn100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 23 Element Oil Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Al,Ba,B
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Al, Ba, B
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.